missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Phenylisoxazol-5-yl)methylamine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 54408-35-4 |
|---|---|
| Molecular Formula | C10H10N2O |
| Molecular Weight (g/mol) | 174.203 |
| InChI Key | AQZLTCXQTOKUAA-UHFFFAOYSA-N |
| Synonym | 3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine |
| PubChem CID | 2764165 |
| IUPAC Name | (3-phenyl-1,2-oxazol-5-yl)methanamine |
| SMILES | C1=CC=C(C=C1)C2=NOC(=C2)CN |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
CC64513CB
|
N/A | 250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC64513DA
|
N/A | 1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC64513DE
|
N/A | 5g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 54408-35-4 | |
| 174.203 | |
| 3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine | |
| (3-phenyl-1,2-oxazol-5-yl)methanamine |
| C10H10N2O | |
| AQZLTCXQTOKUAA-UHFFFAOYSA-N | |
| 2764165 | |
| C1=CC=C(C=C1)C2=NOC(=C2)CN |
Specifications
| 54408-35-4 | |
| C10H10N2O | |
| AQZLTCXQTOKUAA-UHFFFAOYSA-N | |
| (3-phenyl-1,2-oxazol-5-yl)methanamine | |
| 2764165 | |
| 95% |
| 95% | |
| 3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine | |
| C1=CC=C(C=C1)C2=NOC(=C2)CN | |
| 174.203 | |
| 174.2 | |
| (3-phenylisoxazol-5-yl)methylamine |
Safety and Handling
GHS Signal Word: Danger