missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(3-Phenylisoxazol-5-yl)methylamine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC64513CB
| Quantity | 250mg |
|---|---|
| Packaging | Amber Glass Bottle |
Chemical Identifiers
| 54408-35-4 | |
| 174.203 | |
| 3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine | |
| (3-phenyl-1,2-oxazol-5-yl)methanamine |
| C10H10N2O | |
| AQZLTCXQTOKUAA-UHFFFAOYSA-N | |
| 2764165 | |
| C1=CC=C(C=C1)C2=NOC(=C2)CN |
Specifications
| (3-phenylisoxazol-5-yl)methylamine | |
| 95% | |
| C10H10N2O | |
| 3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine | |
| C1=CC=C(C=C1)C2=NOC(=C2)CN | |
| 174.203 | |
| 174.2 |
| 54408-35-4 | |
| Amber Glass Bottle | |
| 250mg | |
| AQZLTCXQTOKUAA-UHFFFAOYSA-N | |
| (3-phenyl-1,2-oxazol-5-yl)methanamine | |
| 2764165 | |
| 95% |
Safety and Handling
GHS Signal Word: Danger