missing translation for 'onlineSavingsMsg'
Learn More

(3-Phenylisoxazol-5-yl)methylamine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC64513DE

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC64513DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

54408-35-4
174.203
3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine
(3-phenyl-1,2-oxazol-5-yl)methanamine
C10H10N2O
AQZLTCXQTOKUAA-UHFFFAOYSA-N
2764165
C1=CC=C(C=C1)C2=NOC(=C2)CN

Specifications

(3-phenylisoxazol-5-yl)methylamine
95%
C10H10N2O
3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine, 3-phenyl-1,2-oxazol-5-yl methanamine, 3-phenylisoxazol-5-yl methylamine, 1-3-phenyl-1,2-oxazol-5-yl methanamine, 5-aminomethyl-3-phenylisoxazole, 1-3-phenylisoxazol-5-yl methanamine, 3-phenyl-5-isoxazolyl methanamine hydrochloride, phenylisoxazolylmethanamine, 3-phenylisoxazol-5-ylmethylamine
C1=CC=C(C=C1)C2=NOC(=C2)CN
174.203
174.2
54408-35-4
Amber Glass Bottle
5g
AQZLTCXQTOKUAA-UHFFFAOYSA-N
(3-phenyl-1,2-oxazol-5-yl)methanamine
2764165
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions