missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenyl-2-propyn-1-ol 97.0+%, TCI America™
Supplier: TCI America P114725G
Specifications
| 3-Phenyl-2-propyn-1-ol | |
| 145°C | |
| MFCD00040914 | |
| 3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol | |
| C1=CC=C(C=C1)C#CCO | |
| 132.162 | |
| 132.16 | |
| Liquid |
| 1504-58-1 | |
| C9H8O | |
| 25 g | |
| NITUNGCLDSFVDL-UHFFFAOYSA-N | |
| 3-phenylprop-2-yn-1-ol | |
| 123115 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 1504-58-1 | |
| 132.162 | |
| NITUNGCLDSFVDL-UHFFFAOYSA-N | |
| 123115 | |
| C1=CC=C(C=C1)C#CCO |
| C9H8O | |
| MFCD00040914 | |
| 3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol | |
| 3-phenylprop-2-yn-1-ol |
Safety and Handling
TSCA : No