CAS RN 1504-58-1
3-Phenyl-2-propyn-1-ol, 98+%
CAS: 1504-58-1 Molecular Formula: C9H8O Molecular Weight (g/mol): 132.162 MDL Number: MFCD00040914 InChI Key: NITUNGCLDSFVDL-UHFFFAOYSA-N Synonym: 3-phenyl-2-propyn-1-ol,2-propyn-1-ol, 3-phenyl,3-phenyl-prop-2-yn-1-ol,1-hydroxymethyl-2-phenylacetylene,1-phenyl-1-propyn-3-ol,pubchem13580,3-phenyl-2-propyn1-ol,3-phenylpropargyl alcohol,acmc-1bs1u,1-phenyl-propyn-3-ol PubChem CID: 123115 IUPAC Name: 3-phenylprop-2-yn-1-ol SMILES: C1=CC=C(C=C1)C#CCO
3-Phenyl-2-propyn-1-ol, 98%
CAS: 1504-58-1 Molecular Formula: C9H8O Molecular Weight (g/mol): 132.16 MDL Number: MFCD00040914 InChI Key: NITUNGCLDSFVDL-UHFFFAOYSA-N Synonym: 3-phenyl-2-propyn-1-ol,2-propyn-1-ol, 3-phenyl,3-phenyl-prop-2-yn-1-ol,1-hydroxymethyl-2-phenylacetylene,1-phenyl-1-propyn-3-ol,pubchem13580,3-phenyl-2-propyn1-ol,3-phenylpropargyl alcohol,acmc-1bs1u,1-phenyl-propyn-3-ol PubChem CID: 123115 IUPAC Name: 3-phenylprop-2-yn-1-ol SMILES: C1=CC=C(C=C1)C#CCO
3-Phenyl-2-propyn-1-ol, 97%
CAS: 1504-58-1 Molecular Formula: C9H8O Molecular Weight (g/mol): 132.162 MDL Number: MFCD00040914 InChI Key: NITUNGCLDSFVDL-UHFFFAOYSA-N Synonym: 3-phenyl-2-propyn-1-ol,2-propyn-1-ol, 3-phenyl,3-phenyl-prop-2-yn-1-ol,1-hydroxymethyl-2-phenylacetylene,1-phenyl-1-propyn-3-ol,pubchem13580,3-phenyl-2-propyn1-ol,3-phenylpropargyl alcohol,acmc-1bs1u,1-phenyl-propyn-3-ol PubChem CID: 123115 IUPAC Name: 3-phenylprop-2-yn-1-ol SMILES: C1=CC=C(C=C1)C#CCO
3-Phenyl-2-propyn-1-ol 97.0+%, TCI America™
CAS: 1504-58-1 Molecular Formula: C9H8O Molecular Weight (g/mol): 132.162 MDL Number: MFCD00040914 InChI Key: NITUNGCLDSFVDL-UHFFFAOYSA-N Synonym: 3-phenyl-2-propyn-1-ol,2-propyn-1-ol, 3-phenyl,3-phenyl-prop-2-yn-1-ol,1-hydroxymethyl-2-phenylacetylene,1-phenyl-1-propyn-3-ol,pubchem13580,3-phenyl-2-propyn1-ol,3-phenylpropargyl alcohol,acmc-1bs1u,1-phenyl-propyn-3-ol PubChem CID: 123115 IUPAC Name: 3-phenylprop-2-yn-1-ol SMILES: C1=CC=C(C=C1)C#CCO