missing translation for 'onlineSavingsMsg'
Learn More

3-Phenyl-2-propyn-1-ol 97.0+%, TCI America™

Catalog No. p-7134974
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11475G 5 g
P114725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P11475G Supplier TCI America Supplier No. P11475G
Only null left

Chemical Identifiers

CAS 1504-58-1
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00040914
InChI Key NITUNGCLDSFVDL-UHFFFAOYSA-N
Synonym 3-phenyl-2-propyn-1-ol, 2-propyn-1-ol, 3-phenyl, 3-phenyl-prop-2-yn-1-ol, 1-hydroxymethyl-2-phenylacetylene, 1-phenyl-1-propyn-3-ol, pubchem13580, 3-phenyl-2-propyn1-ol, 3-phenylpropargyl alcohol, acmc-1bs1u, 1-phenyl-propyn-3-ol
PubChem CID 123115
IUPAC Name 3-phenylprop-2-yn-1-ol
SMILES C1=CC=C(C=C1)C#CCO

Specifications

Boiling Point 145°C
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenyl-2-propyn-1-ol