missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Phenoxypropyl Bromide 95.0+%, TCI America™
Supplier: TCI America P1027250G
Specifications
| 3-Phenoxypropyl Bromide | |
| Yellow | |
| C9H11BrO | |
| 250 g | |
| NIDWUZTTXGJFNN-UHFFFAOYSA-N | |
| (3-bromopropoxy)benzene | |
| 68522 | |
| ≥95.0% (GC) |
| 588-63-6 | |
| 138°C | |
| MFCD00000256 | |
| 3-bromopropoxy benzene, 3-phenoxypropyl bromide, benzene, 3-bromopropoxy, 3-bromopropyl phenyl ether, 1-bromo-3-phenoxypropane, ether, 3-bromopropyl phenyl, gamma-phenoxypropyl bromide, phenoxypropyl bromide, 3-bromo-1-phenoxypropane, .gamma.-phenoxypropyl bromide | |
| BrCCCOC1=CC=CC=C1 | |
| 215.09 | |
| 215.09 | |
| Liquid |
Chemical Identifiers
| 588-63-6 | |
| 215.09 | |
| NIDWUZTTXGJFNN-UHFFFAOYSA-N | |
| 68522 | |
| BrCCCOC1=CC=CC=C1 |
| C9H11BrO | |
| MFCD00000256 | |
| 3-bromopropoxy benzene, 3-phenoxypropyl bromide, benzene, 3-bromopropoxy, 3-bromopropyl phenyl ether, 1-bromo-3-phenoxypropane, ether, 3-bromopropyl phenyl, gamma-phenoxypropyl bromide, phenoxypropyl bromide, 3-bromo-1-phenoxypropane, .gamma.-phenoxypropyl bromide | |
| (3-bromopropoxy)benzene |
Safety and Handling
TSCA : Yes