missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxypropyl Bromide 95.0+%, TCI America™

Catalog No. p-7134908
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P102725G 25 g
P1027250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P102725G Supplier TCI America Supplier No. P102725G
Only null left

Chemical Identifiers

CAS 588-63-6
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD00000256
InChI Key NIDWUZTTXGJFNN-UHFFFAOYSA-N
Synonym 3-bromopropoxy benzene, 3-phenoxypropyl bromide, benzene, 3-bromopropoxy, 3-bromopropyl phenyl ether, 1-bromo-3-phenoxypropane, ether, 3-bromopropyl phenyl, gamma-phenoxypropyl bromide, phenoxypropyl bromide, 3-bromo-1-phenoxypropane, .gamma.-phenoxypropyl bromide
PubChem CID 68522
IUPAC Name (3-bromopropoxy)benzene
SMILES BrCCCOC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 138°C
Quantity 25 g
Formula Weight 215.09
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Phenoxypropyl Bromide