missing translation for 'onlineSavingsMsg'
Learn More

[3-(Phenoxymethyl)phenyl]methanol, 97%, Thermo Scientific™

Catalog No. CC63609DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC63609DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC63609DA Supplier Thermo Scientific Chemicals Supplier No. CC63609DA
Only null left

Chemical Identifiers

CAS 34904-99-9
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD09025916
InChI Key ZKZSJQJXKWKOJY-UHFFFAOYSA-N
Synonym 3-phenoxymethyl phenyl methanol, 3-phenoxymethyl phenyl methan-1-ol
PubChem CID 12399930
IUPAC Name [3-(phenoxymethyl)phenyl]methanol
SMILES C1=CC=C(C=C1)OCC2=CC=CC(=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 214.26
Percent Purity 97%
Chemical Name or Material [3-(Phenoxymethyl)phenyl]methanol
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.