CAS RN 34904-99-9
IUPAC Name:
[3-(phenoxymethyl)phenyl]methanol
Synonyms:
[3-(phenoxymethyl)phenyl]methanol
Molecular Weight (g/mol):
214.26
Molecular Formula:
C14H14O2
InChi Key:
ZKZSJQJXKWKOJY-UHFFFAOYSA-N
SMILES:
OCC1=CC(COC2=CC=CC=C2)=CC=C1
1
–
1
of
1
results
1
[3-(Phenoxymethyl)phenyl]methanol, 97%, Thermo Scientific™
CAS: 34904-99-9 Molecular Formula: C14H14O2 Molecular Weight (g/mol): 214.264 MDL Number: MFCD09025916 InChI Key: ZKZSJQJXKWKOJY-UHFFFAOYSA-N Synonym: 3-phenoxymethyl phenyl methanol,3-phenoxymethyl phenyl methan-1-ol PubChem CID: 12399930 IUPAC Name: [3-(phenoxymethyl)phenyl]methanol SMILES: C1=CC=C(C=C1)OCC2=CC=CC(=C2)CO