missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorure de phénoxybenzoyle, ≥97%, Thermo Scientific™

Numéro de catalogue. CC57102CB
Modifier l'affichage
Click to view available options
Quantité:
250 mg
Emballage:
Bouteille en verre ambrée
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC57102CB 250 mg Bouteille en verre ambrée
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. CC57102CB Fournisseur Thermo Scientific Chemicals Code fournisseur CC57102CB

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 3586-15-0
Formule moléculaire C13H9ClO2
Poids moléculaire (g/mol) 232.663
Numéro MDL MFCD03424712
Clé InChI TTZXIWBOKOZOPL-UHFFFAOYSA-N
Synonyme benzoyl chloride, 3-phenoxy, m-phenoxybenzoyl chloride, m-phenoxy benzoylchoride, 3-phenoxy benzoyl chloride, 3-phenoxy-benzoyl chloride, benzoyl chloride,3-phenoxy
PubChem CID 2760341
Nom de l’IUPAC 3-phénoxybenzoyle chlorure
SOURIRES C1=CC=C(C=C1)OC2=CC=CC(=C2)C(=O)Cl

Spécifications

Nom ou substance chimique 3-Phenoxybenzoyl chloride
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Bouteille en verre ambrée
Quantité 250 mg
Poids de formule 232.67
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.