Learn More
3-Phenoxybenzeneboronic acid, 97+%
CAS: 221006-66-2 | C12H11BO3 | 214.03 g/mol
Supplier: Thermo Scientific Chemicals H3287903
Description
The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.
Solubility
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
| 3-Phenoxybenzeneboronic acid | |
| 138°C to 143°C | |
| MFCD07780272 | |
| 3-phenoxyphenyl boronic acid, 3-phenoxybenzeneboronic acid, boronic acid, 3-phenoxyphenyl, boronic acid, b-3-phenoxyphenyl, acmc-1cmad | |
| LOPQWMNOCSRRSR-UHFFFAOYSA-N | |
| (3-phenoxyphenyl)boronic acid | |
| 10130574 | |
| ≥97% |
| 221006-66-2 | |
| C12H11BO3 | |
| 1 g | |
| Slightly soluble. | |
| OB(O)C1=CC(OC2=CC=CC=C2)=CC=C1 | |
| 214.03 | |
| 214.03 |
Chemical Identifiers
| 221006-66-2 | |
| 214.03 | |
| LOPQWMNOCSRRSR-UHFFFAOYSA-N | |
| 10130574 | |
| OB(O)C1=CC(OC2=CC=CC=C2)=CC=C1 |
| C12H11BO3 | |
| MFCD07780272 | |
| 3-phenoxyphenyl boronic acid, 3-phenoxybenzeneboronic acid, boronic acid, 3-phenoxyphenyl, boronic acid, b-3-phenoxyphenyl, acmc-1cmad | |
| (3-phenoxyphenyl)boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
Recommended Storage : Ambient temperatures
RUO – Research Use Only