missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxybenzeneboronic acid, 97+%

Catalog No. p-7031709
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32879MD 250 mg
AAH3287903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32879MD Supplier Thermo Scientific Chemicals Supplier No. H32879MD

CAS: 221006-66-2 | C12H11BO3 | 214.03 g/mol

The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The extensive use of products containing PBDEs has resulted in the release of 3-Phenoxybenzeneboronic acid.

Solubility
Slightly soluble.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 221006-66-2
Molecular Formula C12H11BO3
Molecular Weight (g/mol) 214.03
MDL Number MFCD07780272
InChI Key LOPQWMNOCSRRSR-UHFFFAOYSA-N
Synonym 3-phenoxyphenyl boronic acid, 3-phenoxybenzeneboronic acid, boronic acid, 3-phenoxyphenyl, boronic acid, b-3-phenoxyphenyl, acmc-1cmad
PubChem CID 10130574
IUPAC Name (3-phenoxyphenyl)boronic acid
SMILES OB(O)C1=CC(OC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 138°C to 143°C
Quantity 250 mg
Solubility Information Slightly soluble.
Formula Weight 214.03
Percent Purity ≥97%
Chemical Name or Material 3-Phenoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Recommended Storage Ambient temperatures

RUO – Research Use Only