missing translation for 'onlineSavingsMsg'
Learn More

3-Phenoxy-1,2-propanediol 95.0+%, TCI America™

Catalog No. p-7135083
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13355G 5 g
P133525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13355G Supplier TCI America Supplier No. P13355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 538-43-2
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004717
InChI Key FNQIYTUXOKTMDM-UHFFFAOYSA-N
Synonym 3-phenoxy-1,2-propanediol, antodyne, phenylglyceryl ether, antodyn, phenol-glycerinaether, phenol glycerol ether, phenol glyceryl ether, 1,2-propanediol, 3-phenoxy, 1-phenoxy-2,3-propanediol, glycerol alpha-monophenyl ether
PubChem CID 10857
IUPAC Name 3-phenoxypropane-1,2-diol
SMILES C1=CC=C(C=C1)OCC(CO)O

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 202°C
Quantity 5 g
Formula Weight 168.19
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Phenoxy-1,2-propanediol
EINECSNumber (3)-0561
RTECSNumber TZ1050000
TSCA No