accessibility menu, dialog, popup

UserName

CAS RN 538-43-2

IUPAC Name: 3-phenoxypropane-1,2-diol
Synonyms: phenyl glycerol
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: FNQIYTUXOKTMDM-UHFFFAOYNA-N
SMILES: OCC(O)COC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results