missing translation for 'onlineSavingsMsg'
Learn More

3-Perylenecarboxaldehyde 95.0+%, TCI America™

Catalog No. P21101G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21101G 1 g
P21105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21101G Supplier TCI America Supplier No. P21101G
Only null left

Chemical Identifiers

CAS 35438-63-2
Molecular Formula C21H12O
Molecular Weight (g/mol) 280.326
MDL Number MFCD00127748
InChI Key IQZOCQOQNWTNOW-UHFFFAOYSA-N
Synonym 3-Formylperylene
PubChem CID 3698916
IUPAC Name perylene-3-carbaldehyde
SMILES C1=CC2=C3C(=C1)C4=C5C(=CC=C4)C(=CC=C5C3=CC=C2)C=O

Specifications

Melting Point 235°C
Quantity 1 g
Formula Weight 280.33
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Perylenecarboxaldehyde