accessibility menu, dialog, popup

UserName

O

CAS RN 35438-63-2

IUPAC Name: perylene-3-carbaldehyde
Synonyms: perylene-3-carbaldehyde
Molecular Weight (g/mol): 280.33
Molecular Formula: C21H12O
InChi Key: IQZOCQOQNWTNOW-UHFFFAOYSA-N
SMILES: O=CC1=C2C=CC=C3C4=CC=CC5=CC=CC(C(C=C1)=C23)=C45
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results