missing translation for 'onlineSavingsMsg'
Learn More

3-Pentadecylphenol 90.0+%, TCI America™

Supplier:  TCI America P1202100G

 View more versions of this product

Catalog No. P1202100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Pentadecylphenol
50°C
C21H36O
100 g
PTFIPECGHSYQNR-UHFFFAOYSA-N
3-pentadecylphenol
68146
≥90.0% (GC)
501-24-6
232°C
MFCD00002310
3-n-pentadecylphenol, hydroginkgol, phenol, 3-pentadecyl, hydrocardanol, cyclogallipharaol, m-pentadecylphenol, tetrahydroanacardol, 3-pentadecyl-phenol, anacardol, tetrahydro, 3-pentadecyl phenol
CCCCCCCCCCCCCCCC1=CC=CC(O)=C1
304.52
304.52
Crystalline Powder

Chemical Identifiers

501-24-6
304.52
PTFIPECGHSYQNR-UHFFFAOYSA-N
68146
CCCCCCCCCCCCCCCC1=CC=CC(O)=C1
C21H36O
MFCD00002310
3-n-pentadecylphenol, hydroginkgol, phenol, 3-pentadecyl, hydrocardanol, cyclogallipharaol, m-pentadecylphenol, tetrahydroanacardol, 3-pentadecyl-phenol, anacardol, tetrahydro, 3-pentadecyl phenol
3-pentadecylphenol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0511

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions