missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Pentadecylphenol 90.0+%, TCI America™
$59.65 - $169.63
Chemical Identifiers
| CAS | 501-24-6 |
|---|---|
| Molecular Formula | C21H36O |
| Molecular Weight (g/mol) | 304.52 |
| MDL Number | MFCD00002310 |
| InChI Key | PTFIPECGHSYQNR-UHFFFAOYSA-N |
| Synonym | 3-n-pentadecylphenol, hydroginkgol, phenol, 3-pentadecyl, hydrocardanol, cyclogallipharaol, m-pentadecylphenol, tetrahydroanacardol, 3-pentadecyl-phenol, anacardol, tetrahydro, 3-pentadecyl phenol |
| PubChem CID | 68146 |
| IUPAC Name | 3-pentadecylphenol |
| SMILES | CCCCCCCCCCCCCCCC1=CC=CC(O)=C1 |
Chemical Identifiers
| 501-24-6 | |
| 304.52 | |
| PTFIPECGHSYQNR-UHFFFAOYSA-N | |
| 68146 | |
| CCCCCCCCCCCCCCCC1=CC=CC(O)=C1 |
| C21H36O | |
| MFCD00002310 | |
| 3-n-pentadecylphenol, hydroginkgol, phenol, 3-pentadecyl, hydrocardanol, cyclogallipharaol, m-pentadecylphenol, tetrahydroanacardol, 3-pentadecyl-phenol, anacardol, tetrahydro, 3-pentadecyl phenol | |
| 3-pentadecylphenol |
Specifications
| 501-24-6 | |
| 232°C | |
| MFCD00002310 | |
| 3-n-pentadecylphenol, hydroginkgol, phenol, 3-pentadecyl, hydrocardanol, cyclogallipharaol, m-pentadecylphenol, tetrahydroanacardol, 3-pentadecyl-phenol, anacardol, tetrahydro, 3-pentadecyl phenol | |
| CCCCCCCCCCCCCCCC1=CC=CC(O)=C1 | |
| 304.52 | |
| 304.52 | |
| Crystalline Powder |
| 50°C | |
| C21H36O | |
| 25 g | |
| PTFIPECGHSYQNR-UHFFFAOYSA-N | |
| 3-pentadecylphenol | |
| 68146 | |
| ≥90.0% (GC) | |
| 3-Pentadecylphenol |
Safety and Handling
EINECSNumber : (3)-0511
TSCA : Yes