missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Octanol 98.0+%, TCI America™
$62.66 - $586.67
Chemical Identifiers
| CAS | 589-98-0 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.23 |
| MDL Number | MFCD00004590 |
| InChI Key | NMRPBPVERJPACX-UHFFFAOYNA-N |
| Synonym | 3-octanol, octanol-3, amyl ethyl carbinol, ethyl-n-amylcarbinol, amylethylcarbinol, ethylamylcarbinol, 1-ethylhexanol, d-n-octanol, ethyl amyl carbinol, s-3-octanol |
| PubChem CID | 11527 |
| ChEBI | CHEBI:80945 |
| IUPAC Name | octan-3-ol |
| SMILES | CCCCCC(O)CC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
O012125ML
|
N/A | 25 mL |
Each for $62.66
|
|
|||||
|
O0121100ML
|
N/A | 100 mL |
Each for $159.49
|
|
|||||
|
O0121500ML
|
N/A | 500 mL |
Each for $586.67
|
|
|||||
Chemical Identifiers
| C8H18O | |
| MFCD00004590 | |
| 3-octanol, octanol-3, amyl ethyl carbinol, ethyl-n-amylcarbinol, amylethylcarbinol, ethylamylcarbinol, 1-ethylhexanol, d-n-octanol, ethyl amyl carbinol, s-3-octanol | |
| CHEBI:80945 | |
| CCCCCC(O)CC |
Specifications
| 589-98-0 | |
| 177°C | |
| MFCD00004590 | |
| NMRPBPVERJPACX-UHFFFAOYNA-N | |
| octan-3-ol | |
| 11527 | |
| 130.23 | |
| Liquid |
| Colorless | |
| C8H18O | |
| 3-octanol, octanol-3, amyl ethyl carbinol, ethyl-n-amylcarbinol, amylethylcarbinol, ethylamylcarbinol, 1-ethylhexanol, d-n-octanol, ethyl amyl carbinol, s-3-octanol | |
| CCCCCC(O)CC | |
| 130.23 | |
| CHEBI:80945 | |
| ≥98.0% (GC) | |
| 3-Octanol |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : RH0855000
TSCA : Yes