missing translation for 'onlineSavingsMsg'
Learn More

3-Octanol 98.0+%, TCI America™

Catalog No. O012125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O012125ML 25 mL
O0121100ML 100 mL
O0121500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. O012125ML Supplier TCI America Supplier No. O012125ML

Chemical Identifiers

CAS 589-98-0
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00004590
InChI Key NMRPBPVERJPACX-UHFFFAOYNA-N
Synonym 3-octanol, octanol-3, amyl ethyl carbinol, ethyl-n-amylcarbinol, amylethylcarbinol, ethylamylcarbinol, 1-ethylhexanol, d-n-octanol, ethyl amyl carbinol, s-3-octanol
PubChem CID 11527
ChEBI CHEBI:80945
IUPAC Name octan-3-ol
SMILES CCCCCC(O)CC

Specifications

Color Colorless
Boiling Point 177°C
Quantity 25 mL
Formula Weight 130.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Octanol
EINECSNumber (2)-0217
RTECSNumber RH0855000
TSCA Yes