missing translation for 'onlineSavingsMsg'
Learn More

3-Octanol 98.0+%, TCI America™

Supplier:  TCI America O0121100ML

Quantity 100 mL

 View more versions of this product

Catalog No. O0121100ML


Only null left

Description

Description

Specifications

Chemical Identifiers

589-98-0
130.23
NMRPBPVERJPACX-UHFFFAOYNA-N
11527
octan-3-ol
C8H18O
MFCD00004590
3-octanol, octanol-3, amyl ethyl carbinol, ethyl-n-amylcarbinol, amylethylcarbinol, ethylamylcarbinol, 1-ethylhexanol, d-n-octanol, ethyl amyl carbinol, s-3-octanol
CHEBI:80945
CCCCCC(O)CC

Specifications

3-Octanol
Colorless
C8H18O
MFCD00004590
NMRPBPVERJPACX-UHFFFAOYNA-N
octan-3-ol
11527
130.23
Liquid
589-98-0
177°C
100 mL
3-octanol, octanol-3, amyl ethyl carbinol, ethyl-n-amylcarbinol, amylethylcarbinol, ethylamylcarbinol, 1-ethylhexanol, d-n-octanol, ethyl amyl carbinol, s-3-octanol
CCCCCC(O)CC
130.23
CHEBI:80945
≥98.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

RTECSNumber : RH0855000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions