missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrothioanisole 98.0+%, TCI America™
$98.79 - $294.52
Chemical Identifiers
| CAS | 2524-76-7 |
|---|---|
| Molecular Formula | C7H7NO2S |
| Molecular Weight (g/mol) | 169.198 |
| MDL Number | MFCD00024501 |
| InChI Key | GBGVXOUUXAIELM-UHFFFAOYSA-N |
| Synonym | 1-(Methylthio)-3-nitrobenzene |
| PubChem CID | 75656 |
| IUPAC Name | 1-methylsulfanyl-3-nitrobenzene |
| SMILES | CSC1=CC=CC(=C1)[N+](=O)[O-] |
Chemical Identifiers
| 2524-76-7 | |
| 169.198 | |
| GBGVXOUUXAIELM-UHFFFAOYSA-N | |
| 75656 | |
| CSC1=CC=CC(=C1)[N+](=O)[O-] |
| C7H7NO2S | |
| MFCD00024501 | |
| 1-(Methylthio)-3-nitrobenzene | |
| 1-methylsulfanyl-3-nitrobenzene |
Specifications
| 2524-76-7 | |
| Yellow | |
| C7H7NO2S | |
| 5 g | |
| GBGVXOUUXAIELM-UHFFFAOYSA-N | |
| 1-methylsulfanyl-3-nitrobenzene | |
| 75656 | |
| ≥98.0% (GC) | |
| 3-Nitrothioanisole |
| 14°C | |
| 92°C | |
| MFCD00024501 | |
| 1-(Methylthio)-3-nitrobenzene | |
| CSC1=CC=CC(=C1)[N+](=O)[O-] | |
| 169.198 | |
| 169.20 | |
| Liquid |
Safety and Handling
TSCA : No