missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrothioanisole 98.0+%, TCI America™
Supplier: TCI America N080025G
Specifications
| 3-Nitrothioanisole | |
| 14°C | |
| 92°C | |
| MFCD00024501 | |
| 1-(Methylthio)-3-nitrobenzene | |
| CSC1=CC=CC(=C1)[N+](=O)[O-] | |
| 169.198 | |
| 169.20 | |
| Liquid |
| 2524-76-7 | |
| Yellow | |
| C7H7NO2S | |
| 25 g | |
| GBGVXOUUXAIELM-UHFFFAOYSA-N | |
| 1-methylsulfanyl-3-nitrobenzene | |
| 75656 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 2524-76-7 | |
| 169.198 | |
| GBGVXOUUXAIELM-UHFFFAOYSA-N | |
| 75656 | |
| CSC1=CC=CC(=C1)[N+](=O)[O-] |
| C7H7NO2S | |
| MFCD00024501 | |
| 1-(Methylthio)-3-nitrobenzene | |
| 1-methylsulfanyl-3-nitrobenzene |
Safety and Handling
TSCA : No