missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrosalicylaldehyde 98.0+%, TCI America™

Catalog No. p-7133985
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08231G 1 g
N08235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08231G Supplier TCI America Supplier No. N08231G
Only null left

Chemical Identifiers

CAS 5274-70-4
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00041874
InChI Key NUGOTBXFVWXVTE-UHFFFAOYSA-N
Synonym 3-nitrosalicylaldehyde, salicylaldehyde, 3-nitro, benzaldehyde, 2-hydroxy-3-nitro, 3-nitro-2-hydroxybenzaldehyde, 3-nitrosalicaldehyde, pubchem7773, 2-hydroxy-nitrobenzaldehyde, acmc-209l1g, 3-nitrosalicylaidehyde, benzaldehyde, 2-hydroxynitro
PubChem CID 78934
IUPAC Name 2-hydroxy-3-nitrobenzaldehyde
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])O)C=O

Specifications

Melting Point 110°C
Quantity 1 g
Formula Weight 167.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrosalicylaldehyde