missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitropyrrole 98.0+%, TCI America™
$439.92 - $1317.89
Chemical Identifiers
| CAS | 5930-94-9 |
|---|---|
| Molecular Formula | C4H4N2O2 |
| Molecular Weight (g/mol) | 112.088 |
| MDL Number | MFCD00059714 |
| InChI Key | LOJNBPNACKZWAI-UHFFFAOYSA-N |
| PubChem CID | 145813 |
| IUPAC Name | 3-nitro-1H-pyrrole |
| SMILES | C1=CNC=C1[N+](=O)[O-] |
Chemical Identifiers
| 5930-94-9 | |
| 112.088 | |
| LOJNBPNACKZWAI-UHFFFAOYSA-N | |
| 3-nitro-1H-pyrrole |
| C4H4N2O2 | |
| MFCD00059714 | |
| 145813 | |
| C1=CNC=C1[N+](=O)[O-] |
Specifications
| 5930-94-9 | |
| White-Yellow | |
| MFCD00059714 | |
| LOJNBPNACKZWAI-UHFFFAOYSA-N | |
| 3-nitro-1H-pyrrole | |
| 145813 | |
| ≥98.0% (GC) | |
| 3-Nitropyrrole |
| 100°C | |
| C4H4N2O2 | |
| 1 g | |
| C1=CNC=C1[N+](=O)[O-] | |
| 112.088 | |
| 112.09 | |
| Crystalline Powder |
Safety and Handling
TSCA : No