accessibility menu, dialog, popup

UserName

CAS RN 5930-94-9

ONONH

CAS RN 5930-94-9

IUPAC Name: 3-nitro-1H-pyrrole
Synonyms: 3-nitro-1H-pyrrole
Molecular Weight (g/mol): 112.09
Molecular Formula: C4H4N2O2
InChi Key: LOJNBPNACKZWAI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CNC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results