missing translation for 'onlineSavingsMsg'
Learn More

3-Nitropyridine 98.0+%, TCI America™

Catalog No. N07715G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N07715G Supplier TCI America Supplier No. N07715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2530-26-9
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.10
MDL Number MFCD00234974
InChI Key QLILRKBRWXALIE-UHFFFAOYSA-N
Synonym 5-nitropyridine, pyridine, 3-nitro, 3-nitro-pyridine, pyridine,3-nitro, pubchem6646, acmc-1cb5f
PubChem CID 137630
IUPAC Name 3-nitropyridine
SMILES [O-][N+](=O)C1=CC=CN=C1

Specifications

Melting Point 40°C
Color Yellow
Boiling Point 216°C
Quantity 5 g
UN Number 2811
Formula Weight 124.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitropyridine