accessibility menu, dialog, popup

UserName

ONON

CAS RN 2530-26-9

IUPAC Name: 3-nitropyridine
Synonyms: 3-nitropyridine
Molecular Weight (g/mol): 124.1
Molecular Formula: C5H4N2O2
InChi Key: QLILRKBRWXALIE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CN=C1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)