missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenylacetic Acid 98.0+%, TCI America™

Catalog No. N04421G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04421G 1 g
N04425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04421G Supplier TCI America Supplier No. N04421G

Chemical Identifiers

CAS 1877-73-2
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD00007278
InChI Key WUKHOVCMWXMOOA-UHFFFAOYSA-N
Synonym 3-nitrophenylacetic acid, 2-3-nitrophenyl acetic acid, 3-nitrophenyl acetic acid, benzeneacetic acid, 3-nitro, m-nitrophenyl acetic acid, m-nitrophenylacetic acid, 3-nitrobenzeneacetic acid, acetic acid, m-nitrophenyl, ccris 2337
PubChem CID 15876
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CC(=O)O

Specifications

Melting Point 119°C
Quantity 1 g
Formula Weight 181.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophenylacetic Acid
RTECSNumber AJ1129900
TSCA No