missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrophenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 98°C |
| Boiling Point | 194°C |
| Quantity | 500 g |
| UN Number | 1663 |
| Formula Weight | 139.11 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Nitrophenol |
Safety and Handling
| EINECSNumber | (3)-0777 |
| RTECSNumber | SM1925000 |
| TSCA | Yes |