CAS RN 554-84-7
3-Nitrophenol, 99%
CAS: 554-84-7 Molecular Formula: C6H5NO3 Molecular Weight (g/mol): 139.11 MDL Number: MFCD00007240 InChI Key: RTZZCYNQPHTPPL-UHFFFAOYSA-N Synonym: m-nitrophenol,3-hydroxynitrobenzene,m-hydroxynitrobenzene,phenol, 3-nitro,phenol, m-nitro,m-nitrofenol,meta-nitrophenol,3-nitro-phenol,m-nitrofenol czech,1-hydroxy-3-nitrobenzene PubChem CID: 11137 ChEBI: CHEBI:34346 IUPAC Name: 3-nitrophenol SMILES: OC1=CC=CC(=C1)[N+]([O-])=O
3-Nitrophenol, 98+%
CAS: 554-84-7 Molecular Formula: C6H5NO3 Molecular Weight (g/mol): 139.11 MDL Number: MFCD00007240 InChI Key: RTZZCYNQPHTPPL-UHFFFAOYSA-N Synonym: m-nitrophenol,3-hydroxynitrobenzene,m-hydroxynitrobenzene,phenol, 3-nitro,phenol, m-nitro,m-nitrofenol,meta-nitrophenol,3-nitro-phenol,m-nitrofenol czech,1-hydroxy-3-nitrobenzene PubChem CID: 11137 ChEBI: CHEBI:34346 IUPAC Name: 3-nitrophenol SMILES: OC1=CC=CC(=C1)[N+]([O-])=O
3-Nitrophenol 98.0+%, TCI America™
CAS: 554-84-7 Molecular Formula: C6H5NO3 Molecular Weight (g/mol): 139.11 MDL Number: MFCD00007240 InChI Key: RTZZCYNQPHTPPL-UHFFFAOYSA-N Synonym: m-nitrophenol,3-hydroxynitrobenzene,m-hydroxynitrobenzene,phenol, 3-nitro,phenol, m-nitro,m-nitrofenol,meta-nitrophenol,3-nitro-phenol,m-nitrofenol czech,1-hydroxy-3-nitrobenzene PubChem CID: 11137 ChEBI: CHEBI:34346 IUPAC Name: 3-nitrophenol SMILES: OC1=CC=CC(=C1)[N+]([O-])=O
m-Nitrophenol, 99.9%, For GC analysis, MP Biomedicals™
CAS: 554-84-7 Molecular Formula: C6H5NO3 Molecular Weight (g/mol): 139.11 MDL Number: MFCD00007240 InChI Key: RTZZCYNQPHTPPL-UHFFFAOYSA-N Synonym: m-nitrophenol,3-hydroxynitrobenzene,m-hydroxynitrobenzene,phenol, 3-nitro,phenol, m-nitro,m-nitrofenol,meta-nitrophenol,3-nitro-phenol,m-nitrofenol czech,1-hydroxy-3-nitrobenzene PubChem CID: 11137 ChEBI: CHEBI:34346 IUPAC Name: 3-nitrophenol SMILES: OC1=CC=CC(=C1)[N+]([O-])=O