missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenol 98.0+%, TCI America™

Catalog No. n021725g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
N021725G 25 g
N0217100G 100 g
N0217500G 500 g
3 options

Catalog No. N021725G

Supplier: TCI America N021725G

Only null left

Chemical Identifiers

CAS 554-84-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007240
InChI Key RTZZCYNQPHTPPL-UHFFFAOYSA-N
Synonym m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene
PubChem CID 11137
ChEBI CHEBI:34346
IUPAC Name 3-nitrophenol
SMILES OC1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 98°C
Boiling Point 194°C
Quantity 25 g
UN Number 1663
Formula Weight 139.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrophenol
EINECSNumber (3)-0777
RTECSNumber SM1925000
TSCA Yes