missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrophenol, 98+%

Catalog No. AAA1531230
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1531230 250 g
AAA1531209 10 g
AAA1531218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1531230 Supplier Thermo Scientific Chemicals Supplier No. A1531230
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 554-84-7 | C6H5NO3 | 139.11 g/mol

3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrophenol is used as indicator in 0.3% Soln in 50% alcohol ph: 6.8 Colorless, 8.6 Yellow. It is used in the synthesis of RAGE inhibitors via the etherification of phenols. Also used in the synthesis of kinase inhibitors against epidermal growth factor receptor T790M mutation.

Solubility
Soluble in water (30 mg/ml), ether, alcohol, acetone, and ethanol (700 mg/ml).

Notes
Store in a tightly closed container. Keep from contact with oxidizing materials. Store in a cool, dry, well-ventilated area away from incompatible substances.

Chemical Identifiers

CAS 554-84-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007240
InChI Key RTZZCYNQPHTPPL-UHFFFAOYSA-N
Synonym m-nitrophenol, 3-hydroxynitrobenzene, m-hydroxynitrobenzene, phenol, 3-nitro, phenol, m-nitro, m-nitrofenol, meta-nitrophenol, 3-nitro-phenol, m-nitrofenol czech, 1-hydroxy-3-nitrobenzene
PubChem CID 11137
ChEBI CHEBI:34346
IUPAC Name 3-nitrophenol
SMILES OC1=CC=CC(=C1)[N+]([O-])=O

Specifications

Melting Point 93°C to 98°C
Density 1.49
Boiling Point 194°C (70 mmHg)
Flash Point >100°C (212°F)
Quantity 250 g
UN Number UN1663
Beilstein 1907946
Merck Index 14,6619
Solubility Information Soluble in water (30mg/ml),ether,alcohol,acetone,and ethanol (700mg/ml).
Formula Weight 139.11
Percent Purity ≥98%
Chemical Name or Material 3-Nitrophenol
Show More Show Less
Hazard Category H302-H315-H318-H373-H500
Hazard Statement GHS H Statement
H318-H373-H302-H312-H335-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P314-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROPHENOLS
EINECSNumber 209-073-5
RTECSNumber SM1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only