missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl Bromide 98.0+%, TCI America™

Catalog No. p-7133898
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N062510G 10 g
N062525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N062510G Supplier TCI America Supplier No. N062510G
Only null left

Chemical Identifiers

CAS 3958-57-4
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007271
InChI Key LNWXALCHPJANMJ-UHFFFAOYSA-N
Synonym 3-nitrobenzyl bromide, 1-bromomethyl-3-nitrobenzene, m-nitrobenzyl bromide, benzene, 1-bromomethyl-3-nitro, alpha-bromo-3-nitrotoluene, m-bromomethyl nitrobenzene, alpha-bromo-m-nitrotoluene, m-nitro-alpha-bromotoluene, 3-nitrobenzylbromide, toluene, alpha-bromo-m-nitro
PubChem CID 77568
IUPAC Name 1-(bromomethyl)-3-nitrobenzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CBr

Specifications

Melting Point 58°C
Boiling Point 115°C
Quantity 10 g
UN Number 3261
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzyl Bromide