accessibility menu, dialog, popup

UserName

ONOBr

CAS RN 3958-57-4

IUPAC Name: 1-(bromomethyl)-3-nitrobenzene
Synonyms: 1-(bromomethyl)-3-nitrobenzene
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: LNWXALCHPJANMJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(CBr)=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)