missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzyl bromide, 98+%

Catalog No. AAA1431022
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1431022 100 g
AAA1431006 5 g
AAA1431014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1431022 Supplier Thermo Scientific Chemicals Supplier No. A1431022
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3958-57-4 | C7H6BrNO2 | 216.034 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3958-57-4
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007271
InChI Key LNWXALCHPJANMJ-UHFFFAOYSA-N
Synonym 3-nitrobenzyl bromide, 1-bromomethyl-3-nitrobenzene, m-nitrobenzyl bromide, benzene, 1-bromomethyl-3-nitro, alpha-bromo-3-nitrotoluene, m-bromomethyl nitrobenzene, alpha-bromo-m-nitrotoluene, m-nitro-alpha-bromotoluene, 3-nitrobenzylbromide, toluene, alpha-bromo-m-nitro
PubChem CID 77568
IUPAC Name 1-(bromomethyl)-3-nitrobenzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])CBr

Specifications

Melting Point 56°C to 59°C
Boiling Point 153°C to 154°C (7 mmHg)
Quantity 100 g
UN Number UN3261
Beilstein 742795
Formula Weight 216.04
Percent Purity ≥98%
Chemical Name or Material 3-Nitrobenzyl bromide
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 223-557-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only