Learn More
3-Nitrobenzoic acid, 99%
CAS: 121-92-6 | C7H5NO4 | 167.12 g/mol
$67.96 - $1126.26
Chemical Identifiers
| CAS | 121-92-6 |
|---|---|
| Molecular Formula | C7H5NO4 |
| Molecular Weight (g/mol) | 167.12 |
| MDL Number | MFCD00007251 |
| InChI Key | AFPHTEQTJZKQAQ-UHFFFAOYSA-N |
| Synonym | 3-nitrobenzoic acid, m-nitrobenzoic acid, benzoic acid, 3-nitro, benzoic acid, m-nitro, meta-nitrobenzoic acid, metanitrobenzoic acid, m-nitrobenzenecarboxylic acid, 3-nitro-benzoic acid, ccris 2335, nitrobenzoic 3-acid |
| PubChem CID | 8497 |
| SMILES | C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1349430
|
Thermo Scientific Chemicals
A1349430 |
250 g |
Each for $67.96
|
|
|||||
|
AAA134940B
|
Thermo Scientific Chemicals
A134940B |
1000 g |
Each for $252.44
|
|
|||||
|
AAA134940I
|
Thermo Scientific Chemicals
A134940I |
5000 g |
Each for $1,126.26
|
|
|||||
Description
3-Nitrobenzoic acid was used to investigated the role of ozone as additional decomposition or finishing reagent in the degradation of o-, m- and p-nitobenzoic acids
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Nitrobenzoic acid was used to investigated the role of ozone as additional decomposition or finishing reagent in the degradation of o-, m- and p-nitobenzoic acids
Solubility
Soluble in water. Insoluble in acetic acid.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 121-92-6 | |
| 167.12 | |
| AFPHTEQTJZKQAQ-UHFFFAOYSA-N | |
| 8497 |
| C7H5NO4 | |
| MFCD00007251 | |
| 3-nitrobenzoic acid, m-nitrobenzoic acid, benzoic acid, 3-nitro, benzoic acid, m-nitro, meta-nitrobenzoic acid, metanitrobenzoic acid, m-nitrobenzenecarboxylic acid, 3-nitro-benzoic acid, ccris 2335, nitrobenzoic 3-acid | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O |
Specifications
| 121-92-6 | |
| 1.494 | |
| MFCD00007251 | |
| 908644 | |
| 3-nitrobenzoic acid, m-nitrobenzoic acid, benzoic acid, 3-nitro, benzoic acid, m-nitro, meta-nitrobenzoic acid, metanitrobenzoic acid, m-nitrobenzenecarboxylic acid, 3-nitro-benzoic acid, ccris 2335, nitrobenzoic 3-acid | |
| AFPHTEQTJZKQAQ-UHFFFAOYSA-N | |
| 167.12 | |
| 167.12 | |
| 3-Nitrobenzoic acid |
| 140°C to 143°C | |
| C7H5NO4 | |
| 250 g | |
| 14,6589 | |
| Soluble in water. Insoluble in acetic acid. | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O | |
| 8497 | |
| 99% |
Safety and Handling
GHS H Statement
H301-H341-H315-H319-H335
Toxic if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 204-508-5
RTECSNumber : DH5000000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only