accessibility menu, dialog, popup

UserName

CAS RN 121-92-6

IUPAC Name: 3-nitrobenzoic acid
Synonyms: 3-nitrobenzoic acid M-nitrobenzoic acid
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: AFPHTEQTJZKQAQ-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (6)
  • (2)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)