missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzoic acid, 99%

Catalog No. AAA1349430
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1349430 250 g
AAA134940B 1000 g
AAA134940I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1349430 Supplier Thermo Scientific Chemicals Supplier No. A1349430
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121-92-6 | C7H5NO4 | 167.12 g/mol

3-Nitrobenzoic acid was used to investigated the role of ozone as additional decomposition or finishing reagent in the degradation of o-, m- and p-nitobenzoic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Nitrobenzoic acid was used to investigated the role of ozone as additional decomposition or finishing reagent in the degradation of o-, m- and p-nitobenzoic acids

Solubility
Soluble in water. Insoluble in acetic acid.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 121-92-6
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007251
InChI Key AFPHTEQTJZKQAQ-UHFFFAOYSA-N
Synonym 3-nitrobenzoic acid, m-nitrobenzoic acid, benzoic acid, 3-nitro, benzoic acid, m-nitro, meta-nitrobenzoic acid, metanitrobenzoic acid, m-nitrobenzenecarboxylic acid, 3-nitro-benzoic acid, ccris 2335, nitrobenzoic 3-acid
PubChem CID 8497
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 140°C to 143°C
Density 1.494
Quantity 250 g
Beilstein 908644
Merck Index 14,6589
Solubility Information Soluble in water. Insoluble in acetic acid.
Formula Weight 167.12
Percent Purity 99%
Chemical Name or Material 3-Nitrobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H341-H315-H319-H335
Toxic if swallowed.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 204-508-5
RTECSNumber DH5000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only