missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrobenzenesulfonamide 98.0+%, TCI America™
Supplier: TCI America N06965G
Specifications
| 3-Nitrobenzenesulfonamide | |
| White-Yellow | |
| MFCD00007935 | |
| benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid | |
| C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-] | |
| 202.184 | |
| 202.18 | |
| Crystalline Powder |
| 121-52-8 | |
| C6H6N2O4S | |
| 5 g | |
| TXTQURPQLVHJRE-UHFFFAOYSA-N | |
| 3-nitrobenzenesulfonamide | |
| 67138 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 121-52-8 | |
| 202.184 | |
| TXTQURPQLVHJRE-UHFFFAOYSA-N | |
| 67138 | |
| C1=CC(=CC(=C1)S(=O)(=O)N)[N+](=O)[O-] |
| C6H6N2O4S | |
| MFCD00007935 | |
| benzenesulfonamide, 3-nitro, m-nitrobenzenesulfonamide, 3-nitrobenzolesulfamide, 3-nitrobenzenesulphonamide, 3-nitrobenzene-1-sulfonamide, m-nitrobenzenesulphonamide, 3-nitro-benzenesulfonamide, benzenesulfonamide, m-nitro, 3-nitrobenzene sulfonamide, m-nitrobenzolsulfonamid | |
| 3-nitrobenzenesulfonamide |
Safety and Handling
EINECSNumber : (3)-2007
TSCA : Yes