accessibility menu, dialog, popup

UserName

NH2SOONOO

CAS RN 121-52-8

IUPAC Name: 3-nitrobenzene-1-sulfonamide
Synonyms: M-nitrobenzenesulfonamide
Molecular Weight (g/mol): 202.18
Molecular Formula: C6H6N2O4S
InChi Key: TXTQURPQLVHJRE-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results