missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitrobenzaldehyde 98.0+%, TCI America™
$77.00 - $350.38
Chemical Identifiers
| CAS | 99-61-6 |
|---|---|
| Molecular Formula | C7H5NO3 |
| Molecular Weight (g/mol) | 151.121 |
| MDL Number | MFCD00007249 |
| InChI Key | ZETIVVHRRQLWFW-UHFFFAOYSA-N |
| Synonym | m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde |
| PubChem CID | 7449 |
| IUPAC Name | 3-nitrobenzaldehyde |
| SMILES | C1=CC(=CC(=C1)[N+](=O)[O-])C=O |
Chemical Identifiers
| 99-61-6 | |
| 151.121 | |
| ZETIVVHRRQLWFW-UHFFFAOYSA-N | |
| 7449 | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C=O |
| C7H5NO3 | |
| MFCD00007249 | |
| m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde | |
| 3-nitrobenzaldehyde |
Specifications
| 99-61-6 | |
| White-Yellow | |
| C7H5NO3 | |
| 25 g | |
| ZETIVVHRRQLWFW-UHFFFAOYSA-N | |
| 3-nitrobenzaldehyde | |
| 7449 | |
| ≥98.0% (GC) | |
| 3-Nitrobenzaldehyde |
| 57°C | |
| 114°C | |
| MFCD00007249 | |
| m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde | |
| C1=CC(=CC(=C1)[N+](=O)[O-])C=O | |
| 151.121 | |
| 151.12 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1181
RTECSNumber : CU7250000
TSCA : Yes