missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzaldehyde 98.0+%, TCI America™

Supplier:  TCI America N0129500G

 View more versions of this product

Catalog No. N0129500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Nitrobenzaldehyde
57°C
114°C
MFCD00007249
m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde
C1=CC(=CC(=C1)[N+](=O)[O-])C=O
151.121
151.12
Crystalline Powder
99-61-6
White-Yellow
C7H5NO3
500 g
ZETIVVHRRQLWFW-UHFFFAOYSA-N
3-nitrobenzaldehyde
7449
≥98.0% (GC)

Chemical Identifiers

99-61-6
151.121
ZETIVVHRRQLWFW-UHFFFAOYSA-N
7449
C1=CC(=CC(=C1)[N+](=O)[O-])C=O
C7H5NO3
MFCD00007249
m-nitrobenzaldehyde, benzaldehyde, 3-nitro, 3-formylnitrobenzene, meta-nitrobenzaldehyde, benzaldehyde, m-nitro, 5-nitrobenzaldehyde, unii-g4o92ko71z, 3-nitro-benzaldehyde, ccris 1784, m-nitro-benzaldehyde
3-nitrobenzaldehyde
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1181

RTECSNumber : CU7250000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions