missing translation for 'onlineSavingsMsg'
Learn More

3'-Nitroacetanilide 98.0+%, TCI America™

Catalog No. N010625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N010625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N010625G Supplier TCI America Supplier No. N010625G

Chemical Identifiers

CAS 122-28-1
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00017015
InChI Key KFTYNYHJHKCRKU-UHFFFAOYSA-N
Synonym n-3-nitrophenyl acetamide, 3'-nitroacetanilide, m-nitroacetanilide, 3-nitroacetanilide, acetamide, n-3-nitrophenyl, n-acetyl-m-nitroaniline, 3-nitro-n-acetylaniline, acetanilide, 3'-nitro, unii-qgh8s22nbp, qgh8s22nbp
PubChem CID 31206
IUPAC Name N-(3-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Melting Point 152°C
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Nitroacetanilide
EINECSNumber (3)-0396
TSCA Yes