accessibility menu, dialog, popup

UserName

CH3OHNNOO

CAS RN 122-28-1

IUPAC Name: N-(3-nitrophenyl)acetamide
Synonyms: M-nitroacetanilide
Molecular Weight (g/mol): 180.16
Molecular Formula: C8H8N2O3
InChi Key: KFTYNYHJHKCRKU-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)