missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitro-p-cresol 98.0+%, TCI America™
$48.13 - $107.80
Chemical Identifiers
| CAS | 2042-14-0 |
|---|---|
| Molecular Formula | C7H7NO3 |
| Molecular Weight (g/mol) | 153.137 |
| MDL Number | MFCD00007244 |
| InChI Key | BQEXDUKMTVYBRK-UHFFFAOYSA-N |
| Synonym | 3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol |
| PubChem CID | 16271 |
| IUPAC Name | 4-methyl-3-nitrophenol |
| SMILES | CC1=C(C=C(C=C1)O)[N+](=O)[O-] |
Chemical Identifiers
| 2042-14-0 | |
| 153.137 | |
| BQEXDUKMTVYBRK-UHFFFAOYSA-N | |
| 16271 | |
| CC1=C(C=C(C=C1)O)[N+](=O)[O-] |
| C7H7NO3 | |
| MFCD00007244 | |
| 3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol | |
| 4-methyl-3-nitrophenol |
Specifications
| 2042-14-0 | |
| C7H7NO3 | |
| 5 g | |
| 3-nitro-4-methylphenol, 3-nitro-p-cresol, 4-methyl-3-nitro-phenol, 4-hydroxy-2-nitrotoluene, 2-nitro-4-hydroxytoluene, phenol, 4-methyl-3-nitro, p-cresol, 3-nitro, unii-4b422hud6h, 4-methyl-5-nitrophenol | |
| CC1=C(C=C(C=C1)O)[N+](=O)[O-] | |
| 153.137 | |
| 153.14 | |
| Crystalline Powder |
| 77°C | |
| MFCD00007244 | |
| 2446 | |
| BQEXDUKMTVYBRK-UHFFFAOYSA-N | |
| 4-methyl-3-nitrophenol | |
| 16271 | |
| ≥98.0% (T) | |
| 3-Nitro-p-cresol |
Safety and Handling
EINECSNumber : (3)-0790
TSCA : No