missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Nitro-o-cresol 98.0+%, TCI America™
$42.87 - $126.76
Chemical Identifiers
| CAS | 5460-31-1 |
|---|---|
| Molecular Formula | C7H7NO3 |
| Molecular Weight (g/mol) | 153.137 |
| MDL Number | MFCD00007241 |
| InChI Key | GAKLFAZBKQGUBO-UHFFFAOYSA-N |
| Synonym | 3-nitro-o-cresol, 2-hydroxy-6-nitrotoluene, phenol, 2-methyl-3-nitro, 2-methyl-3-nitro-phenol, 6-hydroxy-2-nitrotoluene, 3-nitro-2-methyl phenol, nitrocresol, zlchem 214, pubchem2030 |
| PubChem CID | 79579 |
| IUPAC Name | 2-methyl-3-nitrophenol |
| SMILES | CC1=C(C=CC=C1O)[N+](=O)[O-] |
Chemical Identifiers
| 5460-31-1 | |
| 153.137 | |
| GAKLFAZBKQGUBO-UHFFFAOYSA-N | |
| 79579 | |
| CC1=C(C=CC=C1O)[N+](=O)[O-] |
| C7H7NO3 | |
| MFCD00007241 | |
| 3-nitro-o-cresol, 2-hydroxy-6-nitrotoluene, phenol, 2-methyl-3-nitro, 2-methyl-3-nitro-phenol, 6-hydroxy-2-nitrotoluene, 3-nitro-2-methyl phenol, nitrocresol, zlchem 214, pubchem2030 | |
| 2-methyl-3-nitrophenol |
Specifications
| 5460-31-1 | |
| C7H7NO3 | |
| 5 g | |
| 3-nitro-o-cresol, 2-hydroxy-6-nitrotoluene, phenol, 2-methyl-3-nitro, 2-methyl-3-nitro-phenol, 6-hydroxy-2-nitrotoluene, 3-nitro-2-methyl phenol, nitrocresol, zlchem 214, pubchem2030 | |
| CC1=C(C=CC=C1O)[N+](=O)[O-] | |
| 153.137 | |
| 153.14 | |
| Crystalline Powder |
| 152°C | |
| MFCD00007241 | |
| 2446 | |
| GAKLFAZBKQGUBO-UHFFFAOYSA-N | |
| 2-methyl-3-nitrophenol | |
| 79579 | |
| ≥98.0% (GC) | |
| 3-Nitro-o-cresol |
Safety and Handling
EINECSNumber : (3)-0790
TSCA : No