missing translation for 'onlineSavingsMsg'
Learn More

3-Nitro-5-(trifluoromethyl)aniline, 98%

Catalog No. AAH3282309
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3282309 10 g
AAH3282303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3282309 Supplier Thermo Scientific Chemicals Supplier No. H3282309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 401-94-5 | C7H5F3N2O2 | 206.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 401-94-5
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.124
MDL Number MFCD00061229
InChI Key LTVWXWWSCLXXAT-UHFFFAOYSA-N
Synonym 3-nitro-5-trifluoromethyl aniline, 3-amino-5-nitrobenzotrifluoride, 3-amino-5-trifluoromethyl nitrobenzene, 3-nitro-5-trifluoromethyl-phenylamine, 3-nitro-5-trifluoromethyl benzenamine, 5-nitro-3-trifluoromethyl phenylamine, benzenamine, 3-nitro-5-trifluoromethyl, pubchem2558, pubchem2795, ksc915s5r
PubChem CID 2775709
IUPAC Name 3-nitro-5-(trifluoromethyl)aniline
SMILES C1=C(C=C(C=C1N)[N+](=O)[O-])C(F)(F)F

Specifications

Melting Point 77°C to 81°C
Quantity 10 g
UN Number UN2811
Formula Weight 206.13
Percent Purity 98%
Chemical Name or Material 3-Nitro-5-(trifluoromethyl)aniline
Hazard Category H302+H332-H311-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only