missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiopropylamine 98.0+%, TCI America™

Catalog No. p-7132342
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M07545ML 5 mL
M075425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M07545ML Supplier TCI America Supplier No. M07545ML
Only null left

Chemical Identifiers

CAS 4104-45-4
Molecular Formula C4H11NS
Molecular Weight (g/mol) 105.199
MDL Number MFCD00041898
InChI Key KKYSBGWCYXYOHA-UHFFFAOYSA-N
Synonym 3-methylthiopropylamine, 3-methylthio propylamine, 1-propanamine, 3-methylthio, 3-methylthiopropanamine, methylmercaptopropylamine, unii-1iap0cgd5s, 3-methylmercaptopropylamine, 3-methylthio-1-propanamine, 3-methylsulfanyl propan-1-amine, 1iap0cgd5s
PubChem CID 77743
ChEBI CHEBI:16696
IUPAC Name 3-methylsulfanylpropan-1-amine
SMILES CSCCCN

Specifications

Color Yellow
Boiling Point 170°C
Quantity 5 mL
UN Number 2735
Formula Weight 105.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Methylthiopropylamine